Gene Transcripts

Gene Symbol
TRIM17
Entrez Gene ID
51127
Gene Name
tripartite motif containing 17
Gene Aliases
RBCC, RNF16, terf
Chromosome Location
Chr. 1: 228407478 - 228416882 on Build GRCh38
UniGene ID
Hs.121748
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024940.2 NP_001020111.1 4 894
NM_001134855.1 NP_001128327.1 4 1046
NM_016102.3 NP_057186.1 4 930
XM_006711779.2 XP_006711842.1 4 957
XM_011544209.2 XP_011542511.1 4 944
XM_011544210.2 XP_011542512.1 4 1046
XM_011544211.2 XP_011542513.1 4 947
XM_017001418.1 XP_016856907.1 3 1502
XM_017001419.1 XP_016856908.1 4 1046
GenBank mRNA AF156271.1 - 4 653
AK055426.1 - 3 1502
AK301105.1 - 4 850
AK308774.1 - 4 921
AK316379.1 - 4 681
BC033788.1 - 4 694 5165609
BI825739.1 - 4 678
BI831987.1 - 4 595
BM563820.1 - 4 576

View Full Product Details