Gene Transcripts
- Gene Symbol
- TRIM68
- Entrez Gene ID
- 55128
- Gene Name
- tripartite motif containing 68
- Gene Aliases
- GC109, RNF137, SS-56, SS56
- Chromosome Location
- Chr. 11: 4598672 - 4608259 on Build GRCh38
- UniGene ID
- Hs.523438
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001304496.1 |
NP_001291425.1
|
4, 5 |
607 |
|
NM_018073.7 |
NP_060543.5
|
5, 6 |
1090 |
|
GenBank mRNA |
AF360739.1 |
- |
5, 6 |
1043 |
|
AF439153.1 |
- |
4, 5 |
800 |
|
AK001231.1 |
- |
2, 3 |
295 |
|
AK022923.1 |
- |
2, 3 |
260 |
|
AK056523.1 |
- |
5, 6 |
1052 |
|
AK291166.1 |
- |
5, 6 |
1002 |
|
AL832730.1 |
- |
5, 6 |
1040 |
|
AY005802.1 |
- |
4, 5 |
476 |
|
BC075058.2 |
- |
4, 5 |
824 |
30915367 |
BC109063.2 |
- |
5, 6 |
1009 |
40033795 |
View Full Product Details