Gene Transcripts

Gene Symbol
ZNF2
Entrez Gene ID
7549
Gene Name
zinc finger protein 2
Gene Aliases
A1-5, ZNF661, Zfp661
Chromosome Location
Chr. 2: 95165417 - 95184317 on Build GRCh38
UniGene ID
Hs.590916
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017396.2 NP_001017396.1 4 1876
NM_001282398.1 NP_001269327.1 4 1834
NM_001291604.1 NP_001278533.1 3 1762
NM_001291605.1 NP_001278534.1 5 1987
NM_021088.3 NP_066574.2 5 1948
GenBank mRNA AK055072.1 - 5 1928
AK290798.1 - 4 1486
AK295739.1 - 4 1465
AK315829.1 - 4 1481
BC005068.1 - 5 1920 3350363
BC009976.2 - 3 1334
CR749650.1 - 3 1713
X60152.1 - 4 1861

View Full Product Details