Gene Transcripts

Gene Symbol
GSG2
Entrez Gene ID
83903
Gene Name
germ cell associated 2, haspin
Gene Aliases
HASPIN
Chromosome Location
Chr. 17: 3723903 - 3726699 on Build GRCh38
UniGene ID
Hs.534059
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031965.2 NP_114171.2 1 1030
GenBank mRNA AB039834.3 - 1 1027
AF289865.1 - 1 1026
AK292715.1 - 1 1016
AK310546.1 - 1 1030
AK315247.1 - 1 1030
BC016626.1 - 1 1016
BC047457.1 - 1 1011 5298590

View Full Product Details