Gene Transcripts

Gene Symbol
PHF1
Entrez Gene ID
5252
Gene Name
PHD finger protein 1
Gene Aliases
MTF2L2, PCL1, PHF2, TDRD19C, hPHF1
Chromosome Location
Chr. 6: 33410751 - 33416453 on Build GRCh38
UniGene ID
Hs.166204
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002636.4 NP_002627.1 14 2139
NM_024165.2 NP_077084.1 15 2245
NR_027692.1 15 2234
XM_011514662.1 XP_011512964.1 15 2162
XM_011514663.1 XP_011512965.1 15 2159
XM_011514664.1 XP_011512966.1 14 2066
XM_011514665.1 XP_011512967.1 15 2182
XM_011514666.1 XP_011512968.1 15 2179
XM_011514669.1 XP_011512971.1 14 1972
XM_017010939.1 XP_016866428.1 13 1850
GenBank mRNA AF029678.1 - 14 1959
AF052205.1 - 15 2062
AK225913.1 - 14 2247
BC008834.2 - 15 2213 4304525

View Full Product Details