Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001077484.1 | NP_001070952.1 | 17 | 3243 | |
NM_001278387.1 | NP_001265316.1 | 18 | 3705 | ||
NM_001278388.1 | NP_001265317.1 | 16 | 3467 | ||
NM_001278389.1 | NP_001265318.1 | 17 | 2732 | ||
NM_030674.3 | NP_109599.3 | 17 | 3582 | ||
GenBank mRNA | AF247166.1 | - | 17 | 2927 | |
AF271070.1 | - | 17 | 2826 | ||
AK074758.1 | - | 10 | 1999 | ||
AK074797.1 | - | 14 | 2303 | ||
AK074851.1 | - | 15 | 2444 | ||
AK074949.1 | - | 15 | 2490 | ||
AK225778.1 | - | 17 | 2847 | ||
BC010620.1 | - | 17 | 2789 | 3871101 | |
BX647547.1 | - | 18 | 3278 | ||
BX648495.1 | - | 17 | 3170 | ||
BX648676.1 | - | 16 | 3037 |