Gene Transcripts

Gene Symbol
TRIM43
Entrez Gene ID
129868
Gene Name
tripartite motif containing 43
Gene Aliases
TRIM43A
Chromosome Location
Chr. 2: 95592018 - 95599723 on Build GRCh38
UniGene ID
Hs.232026
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138800.1 NP_620155.1 4, 5 882
GenBank mRNA BC015353.1 - 4, 5 882

Gene Symbol
TRIM43B
Entrez Gene ID
653192
Gene Name
tripartite motif containing 43B
Gene Aliases
Chromosome Location
Chr. 2: 95476967 - 95484731 on Build GRCh38
UniGene ID
Hs.723811
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164464.1 NP_001157936.1 4, 5 876
XM_011511669.1 XP_011509971.1 4, 5 820

View Full Product Details