Gene Transcripts

Gene Symbol
IFNLR1
Entrez Gene ID
163702
Gene Name
interferon lambda receptor 1
Gene Aliases
CRF2/12, IFNLR, IL-28R1, IL28RA, LICR2
Chromosome Location
Chr. 1: 24154156 - 24187275 on Build GRCh38
UniGene ID
Hs.221375
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170743.3 NP_734464.1 7 2809
NM_173064.2 NP_775087.1 7 2722
NM_173065.2 NP_775088.1 6 2678
XM_006710394.3 XP_006710457.2 6 2873
XM_017000479.1 XP_016855968.1 6 2838
GenBank mRNA AF439325.1 - 7 2782
AK160364.1 - 7 2785
BC140872.1 - 7 2858 9021740

View Full Product Details