Gene Transcripts

Gene Symbol
SLCO6A1
Entrez Gene ID
133482
Gene Name
solute carrier organic anion transporter family member 6A1
Gene Aliases
CT48, GST, OATP-I, OATP6A1, OATPY
Chromosome Location
Chr. 5: 102371782 - 102502213 on Build GRCh38
UniGene ID
Hs.388874
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF505657.1 - 14 2344
BC034976.2 - 13 2189 4820512

View Full Product Details