Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195643.1 | NP_001182572.1 | 2 | 3302 | |
NM_176815.4 | NP_789785.1 | 2 | 3189 | ||
XM_011512537.2 | XP_011510839.1 | 2 | 3367 | ||
GenBank mRNA | AL832912.1 | - | 2 | 2978 | |
BC045541.1 | - | 2 | 3020 | 5262102 |