Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286668.1 | NP_001273597.1 | 3 | 532 | |
NM_001286669.1 | NP_001273598.1 | 3 | 683 | ||
NM_001286671.1 | NP_001273600.1 | 4 | 804 | ||
NM_001286672.1 | NP_001273601.1 | 4 | 804 | ||
NM_001323890.1 | NP_001310819.1 | 4 | 804 | ||
NM_001323892.1 | NP_001310821.1 | 4 | 804 | ||
NM_001323893.1 | NP_001310822.1 | 3 | 683 | ||
NM_001323899.1 | NP_001310828.1 | 4 | 804 | ||
NM_181806.3 | NP_861522.2 | 4 | 804 | ||
XM_017007740.1 | XP_016863229.1 | 4 | 765 | ||
XM_017007741.1 | XP_016863230.1 | 5 | 880 | ||
XM_017007742.1 | XP_016863231.1 | 2 | 580 | ||
XM_017007743.1 | XP_016863232.1 | 4 | 765 | ||
XM_017007744.1 | XP_016863233.1 | 2 | 580 | ||
GenBank mRNA | AF516672.1 | - | 3 | 447 | |
AK303733.1 | - | 3 | 636 | ||
AK304309.1 | - | 4 | 795 | ||
AK316296.1 | - | 4 | 785 | ||
AY314787.1 | - | 4 | 767 | ||
AY422212.1 | - | 4 | 719 | ||
BC141824.1 | - | 4 | 762 | 40148174 | |
BC142709.1 | - | 4 | 740 | 40148820 | |
BX640635.1 | - | 2 | 331 | ||
BX648853.1 | - | 4 | 796 |