Gene Transcripts

Gene Symbol
TDRD10
Entrez Gene ID
126668
Gene Name
tudor domain containing 10
Gene Aliases
Chromosome Location
Chr. 1: 154502219 - 154548147 on Build GRCh38
UniGene ID
Hs.387671
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098475.1 NP_001091945.1 12 1818
NM_182499.3 NP_872305.3 12 1818
XM_006711156.2 XP_006711219.1 7 1448
XM_011509152.1 XP_011507454.1 13 1207
XM_011509153.1 XP_011507455.1 13 1207
XM_011509154.1 XP_011507456.1 13 1165
XM_011509155.1 XP_011507457.1 13 1207
XM_011509156.1 XP_011507458.1 12 1132
XM_011509157.1 XP_011507459.1 12 1132
XM_011509158.2 XP_011507460.1 8 922
XM_011509159.2 XP_011507461.1 8 913
XM_011509160.2 XP_011507462.1 8 958
XM_011509161.1 XP_011507463.1 8 926
XM_017000255.1 XP_016855744.1 7 852
GenBank mRNA AK302144.1 - 12 1065
AL713777.1 - 7 723
AY358195.1 - 6 587
BC044864.1 - 12 1817 4836747
BC100895.1 - 7 696
BC100896.1 - 7 696
BC100897.2 - 7 696
BC100898.2 - 7 696

View Full Product Details