Gene Transcripts

Gene Symbol
KTN1
Entrez Gene ID
3895
Gene Name
kinectin 1
Gene Aliases
CG1, KNT, MU-RMS-40.19
Chromosome Location
Chr. 14: 55580207 - 55684584 on Build GRCh38
UniGene ID
Hs.509414
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079521.1 NP_001072989.1 6 1197
NM_001079522.1 NP_001072990.1 7 1321
NM_001271014.1 NP_001257943.1 7 1321
NM_004986.3 NP_004977.2 6 1197
NR_073128.1 7 1321
NR_073129.1 7 1321
XM_006720138.2 XP_006720201.1 7 1312
XM_006720139.2 XP_006720202.1 6 1160
XM_006720140.3 XP_006720203.1 6 1160
XM_006720141.2 XP_006720204.1 6 1160
XM_011536751.2 XP_011535053.1 6 1418
XM_011536752.1 XP_011535054.1 6 1160
XM_011536753.1 XP_011535055.1 6 1160
XM_017021278.1 XP_016876767.1 6 7010
XM_017021279.1 XP_016876768.1 6 1160
XM_017021280.1 XP_016876769.1 7 1312
XM_017021281.1 XP_016876770.1 7 7134
XM_017021282.1 XP_016876771.1 7 1315
XM_017021283.1 XP_016876772.1 7 1249
XM_017021284.1 XP_016876773.1 6 1160
XM_017021285.1 XP_016876774.1 7 1292
XM_017021286.1 XP_016876775.1 6 1160
GenBank mRNA AK302797.1 - 7 1275
AY264265.1 - 7 1206
BC050555.1 - 6 1136
BC112337.1 - 7 1204 40068009
BC117132.1 - 6 1138 40125683
BC143720.1 - 6 1138 9052234
D13629.1 - 6 1102
L25616.1 - 5 1059
Z22551.1 - 6 1085

View Full Product Details