Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002172.2 | NP_002163.2 | 1 | 156 | |
GenBank mRNA | BC074956.2 | - | 1 | 156 | |
BC104159.1 | - | 1 | 154 | 40029547 | |
BC104160.1 | - | 1 | 154 | 40029549 | |
V00542.1 | - | 1 | 168 |