Gene Transcripts
- Gene Symbol
- ART3
- Entrez Gene ID
- 419
- Gene Name
- ADP-ribosyltransferase 3
- Gene Aliases
- ARTC3
- Chromosome Location
- Chr. 4: 76011180 - 76112802 on Build GRCh38
- UniGene ID
- Hs.24976
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130016.2 |
NP_001123488.1
|
4, 5 |
952 |
|
NM_001130017.2 |
NP_001123489.1
|
4, 5 |
968 |
|
NM_001179.5 |
NP_001170.2
|
4, 5 |
952 |
|
XM_005262997.1 |
XP_005263054.1
|
3, 4 |
827 |
|
XM_005262999.1 |
XP_005263056.1
|
3, 4 |
827 |
|
XM_005263003.1 |
XP_005263060.1
|
3, 4 |
827 |
|
XM_005263004.1 |
XP_005263061.1
|
3, 4 |
827 |
|
XM_006714220.1 |
XP_006714283.1
|
3, 4 |
827 |
|
XM_011531971.1 |
XP_011530273.1
|
3, 4 |
827 |
|
XM_017008206.1 |
XP_016863695.1
|
3, 4 |
827 |
|
XM_017008207.1 |
XP_016863696.1
|
3, 4 |
827 |
|
XM_017008208.1 |
XP_016863697.1
|
5, 6 |
1340 |
|
XM_017008209.1 |
XP_016863698.1
|
3, 4 |
827 |
|
XM_017008210.1 |
XP_016863699.1
|
5, 6 |
1338 |
|
GenBank mRNA |
AK225860.1 |
- |
4, 5 |
948 |
|
AK295305.1 |
- |
4, 5 |
934 |
|
AK308810.1 |
- |
4, 5 |
934 |
|
AY550336.1 |
- |
2, 3 |
115 |
|
AY550341.1 |
- |
2, 3 |
74 |
|
AY550342.1 |
- |
2, 3 |
74 |
|
AY550344.1 |
- |
2, 3 |
74 |
|
AY550345.1 |
- |
2, 3 |
74 |
|
AY550346.1 |
- |
2, 3 |
74 |
|
BC008397.1 |
- |
4, 5 |
939 |
4244549 |
BC017913.1 |
- |
4, 5 |
939 |
4292261 |
BT007249.1 |
- |
3, 4 |
818 |
|
CR541664.1 |
- |
3, 4 |
818 |
|
U47054.1 |
- |
4, 5 |
848 |
|
View Full Product Details