Gene Transcripts

Gene Symbol
CDK20
Entrez Gene ID
23552
Gene Name
cyclin dependent kinase 20
Gene Aliases
CCRK, CDCH, P42, PNQALRE
Chromosome Location
Chr. 9: 87966441 - 87974780 on Build GRCh38
UniGene ID
Hs.522274
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039803.2 NP_001034892.1 8 2115
NM_001170639.1 NP_001164110.1 7 2024
NM_001170640.1 NP_001164111.1 6 1928
NM_012119.4 NP_036251.2 7 2052
NM_178432.3 NP_848519.1 7 2091
XM_017014561.1 XP_016870050.1 8 2060
XM_017014562.1 XP_016870051.1 9 2139
XM_017014563.1 XP_016870052.1 7 1937
XM_017014564.1 XP_016870053.1 8 2061
GenBank mRNA AF035013.1 - 7 1949
AF113130.1 - 5 3267
AK075325.1 - 7 1844
AK226135.1 - 2 5227
BC002655.2 - 7 1749 3605918
BX647489.1 - 5 3283

View Full Product Details