Gene Transcripts
- Gene Symbol
- SCYL3
- Entrez Gene ID
- 57147
- Gene Name
- SCY1 like pseudokinase 3
- Gene Aliases
- PACE-1, PACE1
- Chromosome Location
- Chr. 1: 169853074 - 169894267 on Build GRCh38
- UniGene ID
- Hs.435560
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020423.6 |
NP_065156.5
|
3, 4 |
582 |
|
NM_181093.3 |
NP_851607.2
|
3, 4 |
582 |
|
XM_006711465.1 |
XP_006711528.1
|
3, 4 |
531 |
|
XM_011509801.1 |
XP_011508103.1
|
3, 4 |
671 |
|
XM_011509802.1 |
XP_011508104.1
|
3, 4 |
531 |
|
XM_011509803.1 |
XP_011508105.1
|
2, 3 |
434 |
|
XM_017001862.1 |
XP_016857351.1
|
3, 4 |
531 |
|
XM_017001863.1 |
XP_016857352.1
|
3, 4 |
671 |
|
XM_017001864.1 |
XP_016857353.1
|
3, 4 |
531 |
|
XM_017001865.1 |
XP_016857354.1
|
3, 4 |
630 |
|
GenBank mRNA |
AF540957.1 |
- |
2, 3 |
412 |
|
AK292507.1 |
- |
3, 4 |
528 |
|
AL117233.1 |
- |
3, 4 |
465 |
|
AY144493.1 |
- |
2, 3 |
412 |
|
BC014662.1 |
- |
3, 4 |
519 |
4852719 |
BX647352.1 |
- |
3, 4 |
671 |
|
View Full Product Details