Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002079.2 | NP_002070.1 | 8 | 1180 | |
GenBank mRNA | AF052153.1 | - | 8 | 1000 | |
AK293483.1 | - | 8 | 902 | ||
AK301916.1 | - | 8 | 1078 | ||
AK312684.1 | - | 8 | 1022 | ||
AY513279.1 | - | 8 | 974 | ||
BC000498.2 | - | 8 | 1000 | 2820849 | |
M37400.1 | - | 8 | 996 |