Gene Transcripts

Gene Symbol
PPID
Entrez Gene ID
5481
Gene Name
peptidylprolyl isomerase D
Gene Aliases
CYP-40, CYPD
Chromosome Location
Chr. 4: 158709127 - 158723400 on Build GRCh38
UniGene ID
Hs.183958
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005038.2 NP_005029.1 2 328
GenBank mRNA AK313929.1 - 2 329
BC014893.1 - 2 315
BC030707.2 - 2 324 4819748
BG699755.1 - 2 333
CR542081.1 - 2 216
L11667.1 - 2 315

View Full Product Details