Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007210.3 | NP_009141.2 | 6 | 1228 | |
XM_005268607.1 | XP_005268664.1 | 6 | 1487 | ||
XM_006719214.2 | XP_006719277.1 | 5 | 1062 | ||
XM_011537819.2 | XP_011536121.1 | 7 | 1443 | ||
XM_011537822.2 | XP_011536124.1 | 6 | 1164 | ||
XM_017018744.1 | XP_016874233.1 | 6 | 1301 | ||
XM_017018745.1 | XP_016874234.1 | 7 | 1286 | ||
GenBank mRNA | AB265820.1 | - | 5 | 1113 | |
AK074658.1 | - | 5 | 1126 | ||
AK296852.1 | - | 7 | 1134 | ||
BC035822.1 | - | 6 | 1231 | 5552059 | |
BC114505.1 | - | 4 | 907 | 40006926 | |
Y08565.1 | - | 4 | 907 |