Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152996.2 | NP_694541.2 | 5 | 1052 | |
XM_006710553.3 | XP_006710616.1 | 6 | 1048 | ||
XM_017000937.1 | XP_016856426.1 | 5 | 4194 | ||
XM_017000940.1 | XP_016856429.1 | 6 | 1311 | ||
XM_017000941.1 | XP_016856430.1 | 5 | 1966 | ||
XM_017000942.1 | XP_016856431.1 | 5 | 1771 | ||
GenBank mRNA | AJ507291.1 | - | 5 | 910 | |
AK091215.1 | - | 5 | 1037 | ||
AK297363.1 | - | 3 | 713 | ||
BC059363.1 | - | 5 | 1023 | 30341233 | |
BX648274.1 | - | 2 | 188 |