Gene Transcripts

Gene Symbol
AKR7L
Entrez Gene ID
246181
Gene Name
aldo-keto reductase family 7 like (gene/pseudogene)
Gene Aliases
AFAR3, AFB1-AR3, AKR7A4
Chromosome Location
Chr. 1: 19265982 - 19274074 on Build GRCh38
UniGene ID
Hs.662342
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040288.1 7 1165
NR_040289.1 5 968
GenBank mRNA AJ278012.1 - 6 951
BC035351.1 - 5 835
CN359627.1 - 1 48

View Full Product Details