Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000420.2 | NP_000411.1 | 4 | 578 | |
XM_005249993.2 | XP_005250050.1 | 4 | 429 | ||
GenBank mRNA | AK314831.1 | - | 4 | 479 | |
BC003135.1 | - | 4 | 578 | 2906155 | |
BC050639.1 | - | 4 | 473 | 5742350 | |
FN998902.1 | - | 3 | 368 | ||
HE572750.1 | - | 3 | 368 | ||
M64934.2 | - | 4 | 488 |