Gene Transcripts
- Gene Symbol
- IDH2
- Entrez Gene ID
- 3418
- Gene Name
- isocitrate dehydrogenase (NADP(+)) 2, mitochondrial
- Gene Aliases
- D2HGA2, ICD-M, IDH, IDHM, IDP, IDPM, mNADP-IDH
- Chromosome Location
- Chr. 15: 90083978 - 90102554 on Build GRCh38
- UniGene ID
- Hs.596461
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289910.1 |
NP_001276839.1
|
6, 7 |
755 |
|
NM_001290114.1 |
NP_001277043.1
|
4, 5 |
730 |
|
NM_002168.3 |
NP_002159.2
|
6, 7 |
988 |
|
GenBank mRNA |
AK294148.1 |
- |
6, 7 |
755 |
|
AK299987.1 |
- |
4, 5 |
644 |
|
AK312627.1 |
- |
6, 7 |
952 |
|
AK316388.1 |
- |
6, 7 |
829 |
|
BC009244.2 |
- |
6, 7 |
890 |
2959540 |
BC071828.1 |
- |
6, 7 |
889 |
6090792 |
X69433.1 |
- |
6, 7 |
910 |
|
View Full Product Details