Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012964.2 | NP_001012982.1 | 4 | 677 | |
NM_001012965.2 | NP_001012983.1 | 3 | 629 | ||
NM_001319948.1 | NP_001306877.1 | 4 | 323 | ||
NM_001319949.1 | NP_001306878.1 | 3 | 520 | ||
NM_002774.3 | NP_002765.1 | 5 | 480 | ||
GenBank mRNA | AF013988.1 | - | 5 | 382 | |
AK314897.1 | - | 5 | 385 | ||
AY279383.1 | - | 4 | 324 | ||
AY318867.1 | - | 4 | 486 | ||
AY318868.1 | - | 3 | 315 | ||
AY318869.1 | - | 4 | 472 | ||
AY318870.1 | - | 3 | 424 | ||
BC015525.2 | - | 4 | 453 | 3852344 | |
BT006852.1 | - | 3 | 236 | ||
D78203.1 | - | 5 | 391 | ||
DQ223012.1 | - | 3 | 328 | ||
U62801.1 | - | 5 | 481 |