Gene Transcripts

Gene Symbol
PSMB3
Entrez Gene ID
5691
Gene Name
proteasome subunit beta 3
Gene Aliases
HC10-II
Chromosome Location
Chr. 17: 38752713 - 38764231 on Build GRCh38
UniGene ID
Hs.82793
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002795.3 NP_002786.2 3 401
NR_104194.1 3 401
GenBank mRNA BC013008.2 - 3 364
CN412852.1 - 3 305
D26598.1 - 3 304

View Full Product Details