Gene Transcripts

Gene Symbol
PCSK7
Entrez Gene ID
9159
Gene Name
proprotein convertase subtilisin/kexin type 7
Gene Aliases
LPC, PC7, PC8, SPC7
Chromosome Location
Chr. 11: 117205071 - 117232525 on Build GRCh38
UniGene ID
Hs.648612
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004716.3 NP_004707.2 10 1876
XM_006718940.3 XP_006719003.1 3 635
XM_017018546.1 XP_016874035.1 5 404
GenBank mRNA AK097207.1 - 2 1051
AK222907.1 - 10 1416
AK312429.1 - 10 1451
BC006357.2 - 10 1394 4125594
BC010696.1 - 10 1446 3868257
BT006870.1 - 8 1245
U33849.1 - 10 1396
U40623.1 - 8 1266

View Full Product Details