Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001276479.1 | NP_001263408.1 | 9 | 1013 | |
NM_001276480.1 | NP_001263409.1 | 8 | 977 | ||
NM_004793.3 | NP_004784.2 | 8 | 1298 | ||
NR_076392.1 | 9 | 996 | |||
XM_011528441.2 | XP_011526743.1 | 8 | 1341 | ||
GenBank mRNA | AK056366.1 | - | 8 | 977 | |
AK096433.1 | - | 9 | 1026 | ||
AK096626.1 | - | 9 | 1013 | ||
AK127867.1 | - | 9 | 1078 | ||
AK298531.1 | - | 9 | 977 | ||
AK309720.1 | - | 9 | 1186 | ||
AL096720.1 | - | 7 | 612 | ||
BC000235.1 | - | 8 | 1177 | 3350958 | |
BC109218.1 | - | 8 | 840 | ||
BC139726.1 | - | 5 | 517 | ||
U02389.1 | - | 8 | 1274 |