Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001308171.1 | NP_001295100.1 | 7 | 460 | |
NM_005925.2 | NP_005916.2 | 7 | 460 | ||
XM_011526013.2 | XP_011524315.1 | 7 | 445 | ||
XM_011526014.2 | XP_011524316.1 | 7 | 445 | ||
GenBank mRNA | AY695931.1 | - | 7 | 471 | |
BC136559.1 | - | 7 | 438 | 9020549 | |
BC144244.1 | - | 7 | 438 | 9052764 | |
X81333.1 | - | 7 | 459 |