Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020361.4 | NP_065094.3 | 1 | 353 | |
XM_011517570.2 | XP_011515872.1 | 1 | 892 | ||
XM_017013646.1 | XP_016869135.1 | 1 | 33 | ||
XM_017013647.1 | XP_016869136.1 | 1 | 1082 | ||
GenBank mRNA | AF221594.1 | - | 1 | 310 | |
AY044833.1 | - | 1 | 311 | ||
BC033684.1 | - | 1 | 313 | 5193403 | |
BI756197.1 | - | 1 | 353 |