Gene Transcripts

Gene Symbol
UBAC2
Entrez Gene ID
337867
Gene Name
UBA domain containing 2
Gene Aliases
PHGDHL1
Chromosome Location
Chr. 13: 99200425 - 99386499 on Build GRCh38
UniGene ID
Hs.508545
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144072.1 NP_001137544.1 7 1183
NM_177967.3 NP_808882.1 5 1097
NR_026644.1 7 1327
XM_006719948.3 XP_006720011.1 8 893
XM_011521082.2 XP_011519384.1 5 953
XM_011521083.2 XP_011519385.1 6 660
XM_011521084.2 XP_011519386.1 7 748
XM_017020553.1 XP_016876042.1 2 3975
GenBank mRNA AF339812.1 - 2 167
AK054563.1 - 5 1098
AK055110.1 - 4 880
AK075419.1 - 7 754
AK075516.1 - 5 524
AK124404.1 - 7 481
AK129861.1 - 7 782
AK290342.1 - 7 1183
AK300908.1 - 7 669
BC063559.1 - 5 542
BC072674.1 - 3 319
BC121138.1 - 5 754
BC121139.1 - 7 926 40120987

View Full Product Details