Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000751.2 | NP_000742.1 | 3 | 279 | |
NM_001311195.1 | NP_001298124.1 | 3 | 279 | ||
NM_001311196.1 | NP_001298125.1 | 3 | 279 | ||
XM_011510524.2 | XP_011508826.1 | 3 | 251 | ||
GenBank mRNA | AK291526.1 | - | 3 | 251 | |
AK296704.1 | - | 3 | 251 | ||
AK304890.1 | - | 3 | 251 | ||
AK315297.1 | - | 3 | 251 | ||
BC093923.1 | - | 3 | 255 | 7939768 | |
BC093925.1 | - | 3 | 255 | 7939770 | |
CD013895.1 | - | 3 | 224 | ||
X55019.1 | - | 3 | 227 |