Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289155.1 | NP_001276084.1 | 2 | 197 | |
NM_001289157.1 | NP_001276086.1 | 2 | 197 | ||
NM_002435.2 | NP_002426.1 | 2 | 197 | ||
XM_011521592.1 | XP_011519894.1 | 2 | 273 | ||
XM_011521593.2 | XP_011519895.1 | 2 | 255 | ||
XM_017022207.1 | XP_016877696.1 | 2 | 277 | ||
XM_017022208.1 | XP_016877697.1 | 2 | 255 | ||
GenBank mRNA | AF504648.1 | - | 2 | 162 | |
AK225149.1 | - | 2 | 160 | ||
AK292374.1 | - | 2 | 150 | ||
AK294030.1 | - | 2 | 166 | ||
BC017351.2 | - | 2 | 146 | 5013257 | |
BC046357.1 | - | 2 | 152 | 5745956 | |
BX648475.1 | - | 2 | 174 | ||
CD706167.1 | - | 2 | 188 | ||
X76057.1 | - | 2 | 139 |