Gene Transcripts

Gene Symbol
POU6F2
Entrez Gene ID
11281
Gene Name
POU class 6 homeobox 2
Gene Aliases
RPF-1, WT5, WTSL
Chromosome Location
Chr. 7: 38977878 - 39468601 on Build GRCh38
UniGene ID
Hs.137106
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166018.1 NP_001159490.1 11 1972
NM_007252.3 NP_009183.3 11 2080
XM_011515113.2 XP_011513415.1 10 2100
GenBank mRNA U91935.1 - 10 1915

View Full Product Details