Gene Transcripts

Gene Symbol
SIX2
Entrez Gene ID
10736
Gene Name
SIX homeobox 2
Gene Aliases
Chromosome Location
Chr. 2: 45005185 - 45009645 on Build GRCh38
UniGene ID
Hs.101937
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016932.4 NP_058628.3 2 1878
XM_005264100.3 XP_005264157.1 2 2126
GenBank mRNA AF332197.1 - 2 1878
BC024033.2 - 2 1858 3027992

View Full Product Details