Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199797.1 | NP_001186726.1 | 3 | 436 | |
NM_002832.3 | NP_002823.3 | 3 | 1282 | ||
NM_080588.2 | NP_542155.1 | 2 | 775 | ||
NR_037663.1 | 2, 3 | 1229 | |||
XM_011509819.1 | XP_011508121.1 | 3 | 617 | ||
XM_011509820.2 | XP_011508122.1 | 3 | 321 | ||
XM_011509821.1 | XP_011508123.1 | 2, 3 | 249 | ||
XM_011509822.2 | XP_011508124.1 | 2, 3 | 120 | ||
XM_011509823.1 | XP_011508125.1 | 3 | 617 | ||
XM_017001935.1 | XP_016857424.1 | 3 | 617 | ||
XM_017001936.1 | XP_016857425.1 | 3 | 617 | ||
XM_017001937.1 | XP_016857426.1 | 3 | 435 | ||
GenBank mRNA | AF394064.1 | - | 1 | 219 | |
AK127214.1 | - | 3 | 1275 | ||
AK225920.1 | - | 3 | 280 | ||
AK301051.1 | - | 2, 3 | 250 | ||
AK301757.1 | - | 3 | 303 | ||
AK301980.1 | - | 3 | 325 | ||
AK302869.1 | - | 3 | 430 | ||
BC001746.1 | - | 2 | 775 | 3543111 | |
BG340453.1 | - | 3 | 317 | ||
BT009848.1 | - | 2 | 186 | ||
D11327.1 | - | 3 | 290 | ||
DC399028.1 | - | 3 | 440 | ||
M64322.1 | - | 3 | 278 |