Gene Transcripts
- Gene Symbol
- DMRT2
- Entrez Gene ID
- 10655
- Gene Name
- doublesex and mab-3 related transcription factor 2
- Gene Aliases
- Chromosome Location
- Chr. 9: 1003237 - 1057554 on Build GRCh38
- UniGene ID
- Hs.59506
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF284223.1 |
- |
3 |
1483 |
|
AF284224.1 |
- |
4 |
1066 |
|
AF284225.1 |
- |
3 |
988 |
|
View Full Product Details