Gene Transcripts
- Gene Symbol
- IRF2
- Entrez Gene ID
- 3660
- Gene Name
- interferon regulatory factor 2
- Gene Aliases
- IRF-2
- Chromosome Location
- Chr. 4: 184387722 - 184474572 on Build GRCh38
- UniGene ID
- Hs.654566
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X15949.1 |
- |
9 |
2113 |
|
View Full Product Details