Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001185009.2 | NP_001171938.1 | 9 | 1808 | |
NM_001185010.2 | NP_001171939.1 | 7 | 1126 | ||
NM_002938.4 | NP_002929.1 | 8 | 1286 | ||
GenBank mRNA | AB000468.1 | - | 8 | 1235 | |
AK128038.1 | - | 8 | 2448 | ||
AK309509.1 | - | 7 | 1054 | ||
AK311314.1 | - | 3 | 614 | ||
BC031935.1 | - | 9 | 1809 | 4824114 | |
U95140.1 | - | 8 | 1208 |