Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001243998.1 | NP_001230927.1 | 3 | 123 | |
NM_001243999.1 | NP_001230928.1 | 3 | 123 | ||
NM_001244000.1 | NP_001230929.1 | 3 | 123 | ||
NM_003121.4 | NP_003112.2 | 3 | 123 | ||
GenBank mRNA | AK225225.1 | - | 3 | 121 | |
AK292644.1 | - | 3 | 125 | ||
AK300639.1 | - | 3 | 121 | ||
BC007921.2 | - | 3 | 102 | 4309499 | |
DA957263.1 | - | 3 | 120 | ||
X66079.1 | - | 3 | 103 | ||
X96998.1 | - | 3 | 103 |