Gene Transcripts
- Gene Symbol
- ZNF143
- Entrez Gene ID
- 7702
- Gene Name
- zinc finger protein 143
- Gene Aliases
- SBF, STAF, pHZ-1
- Chromosome Location
- Chr. 11: 9460965 - 9528524 on Build GRCh38
- UniGene ID
- Hs.523471
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282656.1 |
NP_001269585.1
|
5, 6 |
484 |
|
NM_001282657.1 |
NP_001269586.1
|
4, 5 |
394 |
|
NM_003442.5 |
NP_003433.3
|
5, 6 |
487 |
|
XM_005253121.1 |
XP_005253178.1
|
5, 6 |
435 |
|
XM_005253122.1 |
XP_005253179.1
|
5, 6 |
460 |
|
XM_005253127.1 |
XP_005253184.1
|
6, 7 |
513 |
|
XM_011520349.2 |
XP_011518651.1
|
5, 6 |
1206 |
|
XM_017018252.1 |
XP_016873741.1
|
4, 5 |
371 |
|
XM_017018253.1 |
XP_016873742.1
|
4, 5 |
368 |
|
XM_017018254.1 |
XP_016873743.1
|
5, 6 |
472 |
|
XM_017018255.1 |
XP_016873744.1
|
5, 6 |
562 |
|
XM_017018256.1 |
XP_016873745.1
|
4, 5 |
374 |
|
GenBank mRNA |
AF071771.1 |
- |
2, 3 |
107 |
|
AK291133.1 |
- |
5, 6 |
440 |
|
AK297214.1 |
- |
4, 5 |
339 |
|
AK313330.1 |
- |
5, 6 |
473 |
|
BC020219.1 |
- |
5, 6 |
486 |
4902668 |
BG825506.1 |
- |
5, 6 |
488 |
|
View Full Product Details