Gene Transcripts
- Gene Symbol
- FHL3
- Entrez Gene ID
- 2275
- Gene Name
- four and a half LIM domains 3
- Gene Aliases
- SLIM2
- Chromosome Location
- Chr. 1: 37996765 - 38005515 on Build GRCh38
- UniGene ID
- Hs.57687
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001243878.1 |
NP_001230807.1
|
1, 2 |
170 |
|
NM_004468.4 |
NP_004459.2
|
2, 3 |
346 |
|
GenBank mRNA |
AF133732.1 |
- |
1, 2 |
167 |
|
AK290641.1 |
- |
2, 3 |
346 |
|
AK309179.1 |
- |
2, 3 |
263 |
|
BC001351.2 |
- |
2, 3 |
336 |
2964682 |
BC011697.2 |
- |
2, 3 |
336 |
3532981 |
BC014501.1 |
- |
1, 2 |
157 |
4860329 |
BT007052.1 |
- |
1, 2 |
167 |
|
BX384389.2 |
- |
2, 3 |
402 |
|
CR457425.1 |
- |
1, 2 |
167 |
|
U60116.2 |
- |
1, 2 |
206 |
|
View Full Product Details