Gene Transcripts

Gene Symbol
MN1
Entrez Gene ID
4330
Gene Name
MN1 proto-oncogene, transcriptional regulator
Gene Aliases
MGCR, MGCR1, MGCR1-PEN, dJ353E16.2
Chromosome Location
Chr. 22: 27748277 - 27801498 on Build GRCh38
UniGene ID
Hs.268515
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002430.2 NP_002421.3 2 6971
GenBank mRNA X82209.2 - 2 6970

View Full Product Details