Gene Transcripts

Gene Symbol
ZNF16
Entrez Gene ID
7564
Gene Name
zinc finger protein 16
Gene Aliases
HZF1, KOX9
Chromosome Location
Chr. 8: 144930358 - 144950888 on Build GRCh38
UniGene ID
Hs.493225
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029976.2 NP_001025147.2 4 808
NM_006958.2 NP_008889.2 3 722
XM_005272341.2 XP_005272398.1 3 699
XM_005272342.2 XP_005272399.1 3 807
XM_011517298.1 XP_011515600.1 3 824
GenBank mRNA AF244088.1 - 2 633
AK127625.1 - 3 722
AK226016.1 - 3 692
AL834451.1 - 2 639
BC010996.2 - 3 656 3949487
DQ117529.1 - 4 808

View Full Product Details