Gene Transcripts

Gene Symbol
ZNF10
Entrez Gene ID
7556
Gene Name
zinc finger protein 10
Gene Aliases
KOX1
Chromosome Location
Chr. 12: 133130628 - 133159465 on Build GRCh38
UniGene ID
Hs.573983
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015394.4 NP_056209.2 3 299
GenBank mRNA AK314784.1 - 3 560
AL110188.1 - 1 16
BC024182.2 - 3 300 4823159
X52332.1 - 2 183

View Full Product Details