Gene Transcripts

Gene Symbol
NOVA2
Entrez Gene ID
4858
Gene Name
NOVA alternative splicing regulator 2
Gene Aliases
ANOVA, NOVA3
Chromosome Location
Chr. 19: 45933734 - 45973399 on Build GRCh38
UniGene ID
Hs.375439
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002516.3 NP_002507.1 1, 2 143
XM_017026839.1 XP_016882328.1 1, 2 245
GenBank mRNA AF083898.1 - 1, 2 143
U70477.1 - 1, 2 114

View Full Product Details