Gene Transcripts

Gene Symbol
BTBD1
Entrez Gene ID
53339
Gene Name
BTB domain containing 1
Gene Aliases
C15orf1, NS5ATP8
Chromosome Location
Chr. 15: 83016423 - 83067434 on Build GRCh38
UniGene ID
Hs.459149
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011885.1 NP_001011885.1 4 1022
NM_025238.3 NP_079514.1 4 1022
GenBank mRNA AF257241.1 - 4 901
AF355402.1 - 4 902
AF529369.1 - 4 819
AK000731.1 - 4 886
AK225355.1 - 4 886
AK225362.1 - 4 886
AK294410.1 - 2 274
AK298534.1 - 1 169
AL136853.1 - 4 902
AY364253.1 - 4 819
BC017464.1 - 4 849
BC028097.1 - 4 922 5242418

View Full Product Details