Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001287189.1 | NP_001274118.1 | 4 | 572 | |
NM_001287190.1 | NP_001274119.1 | 4 | 418 | ||
NM_002539.2 | NP_002530.1 | 4 | 680 | ||
GenBank mRNA | AK292352.1 | - | 4 | 553 | |
AK312766.1 | - | 2 | 120 | ||
BC025296.1 | - | 4 | 420 | 5088190 | |
BI463057.1 | - | 4 | 418 | ||
M16650.2 | - | 3 | 206 | ||
X55362.1 | - | 4 | 421 |