Gene Transcripts

Gene Symbol
PDE6G
Entrez Gene ID
5148
Gene Name
phosphodiesterase 6G
Gene Aliases
PDEG, RP57
Chromosome Location
Chr. 17: 81650459 - 81663418 on Build GRCh38
UniGene ID
Hs.654482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002602.3 NP_002593.1 3, 4 327
NR_026872.1 2, 3 122
XM_006721942.3 XP_006722005.1 3, 4 1300
XM_017024734.1 XP_016880223.1 3, 4 602
XM_017024735.1 XP_016880224.1 3, 4 333
XM_017024736.1 XP_016880225.1 3, 4 267
GenBank mRNA BC106884.2 - 3, 4 284
BQ638010.1 - 3, 4 268
BX537414.1 - 3, 4 334
BX647779.1 - 2, 3 116
M36476.1 - 3, 4 284

View Full Product Details