Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003054.4 | NP_003045.2 | 11 | 1180 | |
GenBank mRNA | AK314997.1 | - | 11 | 1133 | |
BC030593.1 | - | 10 | 1438 | ||
BC108927.1 | - | 10 | 1045 | 40025174 | |
BC108928.2 | - | 10 | 1045 | 40025175 | |
L09118.1 | - | 11 | 1122 | ||
L14269.1 | - | 11 | 1050 | ||
L23205.1 | - | 11 | 1130 | ||
X71354.1 | - | 11 | 1080 |